Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AMG 579 | 1227067-61-9 | 99.0% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY1291310MG

Encompass_Preferred

AMG 579 is a potent, selective, and efficacious inhibitor of phosphodiesterase 10A (PDE10A) with an IC50 of 0.1 nM. It has shown a statistically significant reduction of PCP induced behavior in rats.

  • Potent, selective, and efficacious inhibitor of phosphodiesterase 10A (PDE10A)
  • IC50 of 0.1 nM for phosphodiesterase 10A
  • Statistically significant reduction of PCP induced behavior in rats
  • Minimum effective dose of 0.3 mg/kg for efficacy in PCP-LMA model
  • Exhibits a superior oral bioavailability of 72% in dogs

Catalog No. 50-003-39172


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.