Learn More
TARGETMOL CHEMICALS INC AMG 579 5MG
SDP
Catalog No. 502581304
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMG 579 is an efficacious and selective inhibitor of PDE10A (IC50 = 0.1 nM). purity: 99%
Catalog No. 50-258-1304 Supplier TARGETMOL CHEMICALS INC Supplier No. T142175MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMG 579 is an efficacious and selective inhibitor of PDE10A (IC50 = 0.1 nM). purity: 99%