Learn More
Medchemexpress LLC HY-12913 5mg Medchemexpress, AMG 579 CAS:1227067-61-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY129135MG
Encompass_Preferred
Medchemexpress, HY-12913 5mg AMG 579 CAS:1227067-61-9 AMG 579 is a potent, selective, and efficacious inhibitor of phosphodiesterase 10A (PDE10A) with an IC50 of 0.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9798