Learn More
Medchemexpress LLC Amg 7905 (tfa) | 00-00-0 | 99.1% | 597.58 | C27H25F6N5O2S | 5 MG
SDP
Catalog No. 5000263937
Encompass_Preferred
AMG 7905 TFA is the trifluoroacetate salt of AMG 7905, a potent transient receptor potential vanilloid 1 (TRPV1) antagonist used in research. It potentiates proton-enhanced TRPV1 activation while blocking capsaicin-induced channel responses, and has been used to study thermoregulation and TRP channel physiology. The compound is supplied as a high-purity research reagent for in vitro and in vivo studies.
  • Potent TRPV1 antagonist activity.
  • Modulates proton-enhanced channel activation and blocks capsaicin responses.
  • High purity (≈99.1%).
  • Provided as the trifluoroacetate (TFA) salt to aid handling and solubility.
  • Available in small research pack sizes suitable for pilot studies.
  • Intended for laboratory research use only.
Catalog No. 50-002-63937 Supplier Medchemexpress LLC Supplier No. HY113654A5MG
AMG 7905 TFA is the trifluoroacetate salt of AMG 7905, a potent transient receptor potential vanilloid 1 (TRPV1) antagonist used in research. It potentiates proton-enhanced TRPV1 activation while blocking capsaicin-induced channel responses, and has been used to study thermoregulation and TRP channel physiology. The compound is supplied as a high-purity research reagent for in vitro and in vivo studies.
  • Potent TRPV1 antagonist activity.
  • Modulates proton-enhanced channel activation and blocks capsaicin responses.
  • High purity (≈99.1%).
  • Provided as the trifluoroacetate (TFA) salt to aid handling and solubility.
  • Available in small research pack sizes suitable for pilot studies.
  • Intended for laboratory research use only.