Learn More
Medchemexpress LLC AMG 837 calcium hydrate | 1259389-38-2 | 99.0% | 455.45 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13967B100MG
Encompass_Preferred
AMG 837 calcium hydrate is a potent, orally bioavailable, and partial agonist of GPR40/FFA1. It inhibits specific [3H]AMG 837 binding at the human FFA1 receptor with a pIC50 of 8.13. AMG 837 calcium hydrate can enhance insulin secretion and lower glucose levels in rodents. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent, orally bioavailable, and partial agonist of GPR40/FFA1.
  • Inhibits specific [3H]AMG 837 binding at the human FFA1 receptor with a pIC50 of 8.13.
  • Enhances insulin secretion and lowers glucose levels in rodents.
  • Click chemistry reagent with an Alkyne group for CuAAc reactions.
Catalog No. 50-003-49505