Learn More
Medchemexpress LLC AMG 837 calcium hydrate | 1259389-38-2 | 99.0% | C26H21F3O3.1/2Ca.H2O | 10MG
SDP
Catalog No. 5000260188
Encompass_Preferred
AMG 837 calcium hydrate is a research-grade small molecule that functions as a potent, orally bioavailable partial agonist of the GPR40/FFA1 receptor. Supplied as a white to off-white solid, it is used in preclinical studies to potentiate glucose-stimulated insulin secretion and to probe GPR40-mediated signaling pathways.
  • Potent partial agonist of GPR40/FFA1 with nanomolar activity.
  • Purity of 99.0% suitable for in vitro and preclinical applications.
  • Solid, white to off-white appearance for easy handling and storage.
  • Stable under recommended storage conditions for long-term use.
  • Useful for studies of insulin secretion and metabolic disease mechanisms.
Catalog No. 50-002-60188 Supplier Medchemexpress LLC Supplier No. HY13967B10MG
AMG 837 calcium hydrate is a research-grade small molecule that functions as a potent, orally bioavailable partial agonist of the GPR40/FFA1 receptor. Supplied as a white to off-white solid, it is used in preclinical studies to potentiate glucose-stimulated insulin secretion and to probe GPR40-mediated signaling pathways.
  • Potent partial agonist of GPR40/FFA1 with nanomolar activity.
  • Purity of 99.0% suitable for in vitro and preclinical applications.
  • Solid, white to off-white appearance for easy handling and storage.
  • Stable under recommended storage conditions for long-term use.
  • Useful for studies of insulin secretion and metabolic disease mechanisms.