Learn More
Apexbio Technology LLC AMG-900 945595-80-2 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A41195.1
Encompass_Preferred
AMG-900 (CAS 945595-80-2) is a small-molecule inhibitor targeting Aurora kinases a group of serine/threonine kinases crucial for mitosis regulation It exhibits potent ATP-competitive inhibitory activity against all three Aurora kinase isoforms resulting in suppression of Aurora-A and Aurora-B autophosphorylation events AMG-900 induces disruption of cellular mitotic processes leading to cell cycle arrest and subsequent apoptotic cell death Preclinically AMG-900 demonstrates activity across multiple tumor cell lines including multidrug-resistant and taxane-resistant variants and inhibits tumor growth in mouse xenograft models suggesting utility for cancer research
Catalog No. 50-000-61540