Learn More
Medchemexpress LLC HY-15889 5mg Medchemexpress, AMG 925 CAS:1401033-86-0 Purity:>98%
SDP
Catalog No. 502081817
Encompass_Preferred
Medchemexpress, HY-15889 5mg AMG 925 CAS:1401033-86-0 AMG 925 is a potent, selective, and orally available FLT3/CDK4 dual inhibitor with IC50s of 2±1 nM and 3±1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1817 Supplier Medchemexpress LLC Supplier No. HY158895MG
Medchemexpress, HY-15889 5mg AMG 925 CAS:1401033-86-0 AMG 925 is a potent, selective, and orally available FLT3/CDK4 dual inhibitor with IC50s of 2±1 nM and 3±1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.