Learn More
Enzo Life Sciences AMG-9810 (10mg). CAS: 545395-94-6
SDP
Catalog No. 502010371
Encompass_Preferred
AMG-9810 is an antagonist of capsaicin activation of TRPV1 (IC50=24.5 nM) as well as activation by acid, heat, and endogenous ligands. Purity: ≥98%. Solubility: Soluble in DMSO (>25mg/ml); ethanol (20mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0371 Supplier Enzo Life Sciences Supplier No. BMLCA2320010
AMG-9810 is an antagonist of capsaicin activation of TRPV1 (IC50=24.5 nM) as well as activation by acid, heat, and endogenous ligands. Purity: ≥98%. Solubility: Soluble in DMSO (>25mg/ml); ethanol (20mg/ml). Long Term Storage: Ambient.