Learn More
Medchemexpress LLC Amg perk 44 | 1883548-84-2 | 99.7% | 100 MG
SDP
Catalog No. 5000338879
Encompass_Preferred
This compound is an orally active and highly selective inhibitor of PERK, demonstrating potent activity with an IC50 of 6 nM. It exhibits significant selectivity, showing 1000-fold preference over GCN2 and 160-fold over B-Raf. Furthermore, this agent has been shown to induce autophagy.
  • Exhibits high potency against its target with a low nanomolar IC50.
  • Demonstrates remarkable selectivity compared to other related kinases.
  • Induces autophagy, suggesting potential in related cellular processes.
  • Effective when administered orally.
  • Robustly inhibits target autophosphorylation in living systems.
  • Maintains sustained target inhibition in vivo for up to 24 hours at certain dosages.
Catalog No. 50-003-38879 Supplier Medchemexpress LLC Supplier No. HY12661A100MG
This compound is an orally active and highly selective inhibitor of PERK, demonstrating potent activity with an IC50 of 6 nM. It exhibits significant selectivity, showing 1000-fold preference over GCN2 and 160-fold over B-Raf. Furthermore, this agent has been shown to induce autophagy.
  • Exhibits high potency against its target with a low nanomolar IC50.
  • Demonstrates remarkable selectivity compared to other related kinases.
  • Induces autophagy, suggesting potential in related cellular processes.
  • Effective when administered orally.
  • Robustly inhibits target autophosphorylation in living systems.
  • Maintains sustained target inhibition in vivo for up to 24 hours at certain dosages.