Learn More
Medchemexpress LLC HY-117103 5mg , AMG131 CAS:315224-26-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1171035MG
Encompass_Preferred
HY-117103 5mg AMG131 CAS: 315224-26-1 AMG131 (INT131), a potent and highly selective PPARγ partial agonist, binds to PPARγ and displaces Rosiglitazone with a Ki of ~10 nM. AMG131 can be used for research of type-2 diabetes mellitus (T2DM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1311