Learn More
Selleck Chemical LLC Sotorasib (AMG510) 25mg 2296729-00-3
SDP
Catalog No. NC1700252
Encompass_Preferred
Sotorasib (AMG510) is a potent KRAS G12C covalent inhibitor with potential antineoplastic activity.This AMG510 is a chiral compound. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1700252 Supplier Selleck Chemical LLC Supplier No. S883025MG
Sotorasib (AMG510) is a potent KRAS G12C covalent inhibitor with potential antineoplastic activity.This AMG510 is a chiral compound. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.