Learn More
TARGETMOL CHEMICALS INC AMG9810 25MG
SDP
Catalog No. 502576337
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMG 9810 is a competitive antagonist of capsaicin activation of the vanilloid receptor 1 (TRPV1) (IC50s = 24.5 and 85.6 nM for human and rat TRPV1 respectively). purity: 98%
Catalog No. 50-257-6337 Supplier TARGETMOL CHEMICALS INC Supplier No. T718925MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMG 9810 is a competitive antagonist of capsaicin activation of the vanilloid receptor 1 (TRPV1) (IC50s = 24.5 and 85.6 nM for human and rat TRPV1 respectively). purity: 98%