Learn More
Selleck Chemical LLC AMI-1 (free acid) S5445-25mg
SDP
Catalog No. 503108603
Encompass_Preferred
AMI-1 is a potent and specific Histone Methyltransferase (HMT) inhibitor with IC50 of 3 0 M and 8 8 M for yeast Hmt1p and human PRMT1 respectively
Catalog No. 50-310-8603 Supplier Selleck Chemical LLC Supplier No. S544525MG
AMI-1 is a potent and specific Histone Methyltransferase (HMT) inhibitor with IC50 of 3 0 M and 8 8 M for yeast Hmt1p and human PRMT1 respectively