Learn More
Selleck Chemical LLC AMI-1
SDP
Catalog No. 5000001632
Encompass_Preferred
AMI-1 is a potent and specific Histone Methyltransferase (HMT) inhibitor with IC50 of 3 0 M and 8 8 M for yeast Hmt1p and human PRMT1 respectively
Catalog No. 50-000-01632 Supplier Selleck Chemical LLC Supplier No. S788410MM/1ML
AMI-1 is a potent and specific Histone Methyltransferase (HMT) inhibitor with IC50 of 3 0 M and 8 8 M for yeast Hmt1p and human PRMT1 respectively