Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ami-1 | 20324-87-2 | 98.0% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY1896210MG

Encompass_Preferred

AMI-1 is a cell-permeable, reversible small-molecule inhibitor of protein arginine N-methyltransferases (PRMTs). It blocks peptide-substrate binding and is used in biochemical and cellular studies to probe arginine methylation and PRMT function.

  • Purity: 98.0%.
  • Molecular weight: 548.45 g·mol⁻1.
  • CAS number: 20324-87-2.
  • Chemical formula: C21H14N2Na2O9S2.
  • Pack size: 10 MG.
  • Storage: sealed, away from moisture; in solution, -80°C up to 2 years or -20°C up to 1 year.
  • Targets: inhibits PRMT1, PRMT3, PRMT4, PRMT6, PRMT5, and yeast Hmt1p.

Catalog No. 50-002-94754


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.