Learn More
Medchemexpress LLC AMI-1 | 20324-87-2 | 98.0% | 548.45 | 25 MG
SDP
Catalog No. 5000385046
Encompass_Preferred
AMI-1 is a potent, cell-permeable, and reversible inhibitor of protein arginine N-methyltransferases (PRMTs), with IC50s of 8.8 μM for human PRMT1 and 3.0 μM for yeast-Hmt1p. It inhibits PRMTs by blocking peptide-substrate binding.
  • Potent, cell-permeable, and reversible inhibitor
  • Inhibits in vitro methylation reactions performed by PRMT1, -3, -4, -6, and Hmt1p
  • Inhibits both type I and type II PRMTs
  • Specifically inhibits arginine methyltransferase activity
  • Does not compete for the AdoMet binding site
  • Inhibits methylation of GFP-Npl3 and cellular proteins
Catalog No. 50-003-85046 Supplier Medchemexpress LLC Supplier No. 25MGHY18962
AMI-1 is a potent, cell-permeable, and reversible inhibitor of protein arginine N-methyltransferases (PRMTs), with IC50s of 8.8 μM for human PRMT1 and 3.0 μM for yeast-Hmt1p. It inhibits PRMTs by blocking peptide-substrate binding.
  • Potent, cell-permeable, and reversible inhibitor
  • Inhibits in vitro methylation reactions performed by PRMT1, -3, -4, -6, and Hmt1p
  • Inhibits both type I and type II PRMTs
  • Specifically inhibits arginine methyltransferase activity
  • Does not compete for the AdoMet binding site
  • Inhibits methylation of GFP-Npl3 and cellular proteins