Learn More
TARGETMOL CHEMICALS INC AMI-1 25MG
Supplier: TARGETMOL CHEMICALS INC T235225MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMI-1 is an effective and selective Histone Methyltransferase (HMT) inhibitor (IC50 3.0/8.8 uM for yeast Hmt1p and human PRMT1). purity: 99%