Learn More
AdipoGen AMI-1 (25 mg)
SDP
Supplier:  AdipoGen AGCR10038M025
Encompass_Preferred
Chemical. CAS: 20324-87-2134-47-4 (free base). Formula: C21H12N2O9S2Na4. MW: 592.4. Potent, cell permeable and specific protein arginine N-methyltransferase 1 (PRMT1) inhibitor (IC50 = 8.8 µM). Does not compete for the AdoMet (S-adenosyl-L-methionine; SAM) binding site. Blocks peptide substrate binding. HIV-1 RT polymerase inhibitor (IC50 = 5.0 µM).
Catalog No. 50-205-3794