Learn More
Medchemexpress LLC AMI-1 free acid | 134-47-4 | 99.5% | 504.49 | 1 ML
SDP
Catalog No. 5000385190
Encompass_Preferred
AMI-1 free acid is a potent, cell-permeable, and reversible inhibitor of protein arginine N-methyltransferases (PRMTs), with IC50s of 8.8 μM for human PRMT1 and 3.0 μM for yeast-Hmt1p. It exerts its inhibitory effects on PRMTs by blocking peptide-substrate binding.
  • Potent, cell-permeable, and reversible inhibitor of PRMTs.
  • Inhibits type I PRMTs and type II PRMT5.
  • Specifically inhibits arginine methyltransferase activity, not lysine, and does not compete for the AdoMet binding site.
  • Inhibits methylation of GFP-Npl3 and cellular proteins.
  • Inhibits the cell viability of sarcoma in S180 and U2OS cells in a time- and dose-dependent manner in vitro.
  • Reduces S180 cell viability through the induction of cell apoptosis.
  • Inhibits S180 viability in vivo.
  • Downregulates PRMT5 and decreases levels of H4R3me2s and H3R8me2s in a tumor xenograft model.
Catalog No. 50-003-85190 Supplier Medchemexpress LLC Supplier No. HY18962A10MM/1ML
AMI-1 free acid is a potent, cell-permeable, and reversible inhibitor of protein arginine N-methyltransferases (PRMTs), with IC50s of 8.8 μM for human PRMT1 and 3.0 μM for yeast-Hmt1p. It exerts its inhibitory effects on PRMTs by blocking peptide-substrate binding.
  • Potent, cell-permeable, and reversible inhibitor of PRMTs.
  • Inhibits type I PRMTs and type II PRMT5.
  • Specifically inhibits arginine methyltransferase activity, not lysine, and does not compete for the AdoMet binding site.
  • Inhibits methylation of GFP-Npl3 and cellular proteins.
  • Inhibits the cell viability of sarcoma in S180 and U2OS cells in a time- and dose-dependent manner in vitro.
  • Reduces S180 cell viability through the induction of cell apoptosis.
  • Inhibits S180 viability in vivo.
  • Downregulates PRMT5 and decreases levels of H4R3me2s and H3R8me2s in a tumor xenograft model.