Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amibegron hydrochloride | 121524-09-2 | MFCD00886563 | >98.0% | 440.36 g/mol | C22H27Cl2NO4 | 1 MG
SDP

Catalog No. 5000247276
Encompass_Preferred

Amibegron hydrochloride is a selective β3-adrenoceptor agonist used in preclinical research. It exhibits an EC50 of 3.5 nM for β-adrenoceptors in rat colon and has reported anxiolytic and antidepressant activity. The compound is supplied as a purified solid for laboratory assays and bioactivity studies; consult the manufacturer's datasheet for handling and storage conditions.

  • Molecular formula: C22H27Cl2NO4.
  • Molecular weight: 440.36 g/mol.
  • CAS number: 121524-09-2.
  • Purity: >98.0% (HPLC).
  • Target: β3-adrenoceptor (adrenergic receptor).
  • Typical package sizes: 1 mg, 5 mg, 10 mg.
  • Storage: -20°C sealed; in solution -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-47276 Supplier Medchemexpress LLC Supplier No. HY1032071MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amibegron hydrochloride is a selective β3-adrenoceptor agonist used in preclinical research. It exhibits an EC50 of 3.5 nM for β-adrenoceptors in rat colon and has reported anxiolytic and antidepressant activity. The compound is supplied as a purified solid for laboratory assays and bioactivity studies; consult the manufacturer's datasheet for handling and storage conditions.

  • Molecular formula: C22H27Cl2NO4.
  • Molecular weight: 440.36 g/mol.
  • CAS number: 121524-09-2.
  • Purity: >98.0% (HPLC).
  • Target: β3-adrenoceptor (adrenergic receptor).
  • Typical package sizes: 1 mg, 5 mg, 10 mg.
  • Storage: -20°C sealed; in solution -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.