Learn More
Medchemexpress LLC Amicarbazone | 129909-90-6 | 98.3% | 241.29 | 250 MG
SDP
Catalog No. 5000384785
Encompass_Preferred
Amicarbazone is a potent herbicide that acts as an inhibitor of photosynthetic electron transport. It functions by binding to the Qb domain of photosystem II (PSII) and is utilized for broad-spectrum weed control activity. This compound is intended for research use only.
  • Potent inhibitor of photosynthetic electron transport
  • Binds to the Qb domain of photosystem II (PSII)
  • Provides broad-spectrum weed control activity
  • Effective in reducing sensitivity in certain annual bluegrass populations
  • Offers complete annual bluegrass control and full quantum yield reduction
Catalog No. 50-003-84785 Supplier Medchemexpress LLC Supplier No. 250MGHY17513
Amicarbazone is a potent herbicide that acts as an inhibitor of photosynthetic electron transport. It functions by binding to the Qb domain of photosystem II (PSII) and is utilized for broad-spectrum weed control activity. This compound is intended for research use only.
  • Potent inhibitor of photosynthetic electron transport
  • Binds to the Qb domain of photosystem II (PSII)
  • Provides broad-spectrum weed control activity
  • Effective in reducing sensitivity in certain annual bluegrass populations
  • Offers complete annual bluegrass control and full quantum yield reduction