Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Amicon™ Pro Affinity concentration Kit GST with Amicon™ Ultra-0.5 Device

Catalog No. ACK5100GS Shop All MilliporeSigma Products
Change view
Click to view available options
MWCO:
100 kDa
30 kDa
50 kDa
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO
ACK5100GS 100 kDa
ACK5050GS 50 kDa
ACK5030GS 30 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ACK5100GS Supplier MilliporeSigma™ Supplier No. ACK5100GS

missing translation for 'validMainframeMsg'

A Centrifugal Tool for purifying and concentrating proteins.

This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/ml agarose), 125 mg Glutathione (reduced, free acid), 25ml GST Bind/Wash Buffer (10X), 5mL GST Reconstitution buffer (10X)

Order Info

Reagents available. (See Cat. No. ACR5000NT)

TRUSTED_SUSTAINABILITY

Specifications

Color Clear with Pink Cap
Material (Membrane) Ultracel Low-binding Regenerated Cellulose
MWCO 100 kDa
Filtration Area 1 cm2
For Use With (Application) Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment
Housing Material Polypropylene
Includes 12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 100 kd, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/mL agarose), 125mg Glutathione (reduced, free acid), 25mL GST Bind/Wash Buffer (10x), 5mL GST Reconstitution buffer (10x)
Max. Force 4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin
Quantity 12/Pk.
Type Amicon Pro Affinity Concentration Kit GST with 100 kd Amicon Ultra-0.5 Device
Volume (Metric) 10 mL
Length (English) 4.7 in.
Length (Metric) 12 cm
Diameter (English) 1.2 in.
Diameter (Metric) 29.7 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.