Learn More
Cayman Chemical AmIdepsIn A 2.5mg
SDP
Catalog No. 502980174
Encompass_Preferred
A fungal metabolite that inhibits DGAT activity, preventing triacylglycerol formation (IC50s = 10.2 and 15.5 µM in rat liver microsomes and Raji cells, respectively
Catalog No. 50-298-0174 Supplier Cayman Chemical Supplier No. 177742.5 MG
A fungal metabolite that inhibits DGAT activity, preventing triacylglycerol formation (IC50s = 10.2 and 15.5 µM in rat liver microsomes and Raji cells, respectively