Learn More
Cayman Chemical AmIdepsIn D 2.5mg
SDP
Catalog No. 502979798
Encompass_Preferred
A fungal metabolite that inhibits DGAT activity, preventing triacylglycerol formation (IC50s = 17.5 and 2.8 µM in rat liver microsomes and Raji cells, respectively)
Catalog No. 50-297-9798 Supplier Cayman Chemical Supplier No. 177752.5 MG
A fungal metabolite that inhibits DGAT activity, preventing triacylglycerol formation (IC50s = 17.5 and 2.8 µM in rat liver microsomes and Raji cells, respectively)