Learn More
AdipoGen Amidepsine A (1 mg)
SDP
Supplier:  AdipoGen AGCN20109M001
Encompass_Preferred
Chemical. CAS: 169181-28-6. Formula: C29H29NO11. MW: 567.6. Isolated from Humcola sp. FO-2942. Diacylglycerol acyltransferase (DGAT) inhibitor. Excessive accumulation of triacetylglycerol produced by DGAT may cause fatty liver, obesity and hypertriglyceridemia, which may lead to atherosclerosis, diabetes and metabolic disorders.
Catalog No. 50-205-3468