Learn More
AdipoGen Amidepsine D (2.5 mg)
SDP
Supplier:  AdipoGen AGCN20110MM25
Encompass_Preferred
Chemical. CAS: 79786-34-8. Formula: C26H24O10. MW: 496.5. Isolated from Humcola sp. FO-2942. Diacylglycerol acyltransferase (DGAT) inhibitor. Excessive accumulation of triacetylglycerol produced by DGAT may cause fatty liver, obesity and hypertriglyceridemia, which may lead to atherosclerosis, diabetes and metabolic disorders.
Catalog No. 50-205-3471