Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amido black 10 B (C.I.20470), For Electrophoresis, Certistain™, MilliporeSigma™

Catalog No. M1011670025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1011670025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1011670025 Supplier MilliporeSigma Supplier No. 1.01167.0025

missing translation for 'validMainframeMsg'

For electrophoresis
TRUSTED_SUSTAINABILITY

Specifications

CAS 1064-48-8
Molecular Formula C22H14N6Na2O9S2
Synonym Black acid 1, Naphthol blue black
InChI Key HKBVRFLHNUEVRO-UHFFFAOYSA-L
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
Molecular Weight (g/mol) 616.487
PubChem CID 44134531
Formula Weight 616.49
Odor Odorless
Packaging Glass bottle
Color Dark Brown
Quantity 25 g
Physical Form Solid
Chemical Name or Material Amido black 10 B (C.I.20470)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.