Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amido Black 10B, Fisher BioReagents

Catalog No. BP12410 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP124-10 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP124-10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Amido Black 10B is a general stain for proteins and nucleic acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1064-48-8
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.487
InChI Key HKBVRFLHNUEVRO-UHFFFAOYSA-L
Synonym Naphthol Blue Black
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Extinction Coefficient 450g(-1) cm(-1) min.
Identification Pass Test
Lambda Max 616 to 620nm
Formula Weight 616.5
Loss on Drying 10% max.
Packaging Glass Bottle
Color Index No. 20470
Color Brown
pH 9.0
Quantity 10 g
Physical Form Powder
Chemical Name or Material Amido Black 10B
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.