Learn More
Mopec Amido Black
SDP
Catalog No. 22039027 Shop All Mopec Products
Change view
Click to view available options
Quantity:
25 g
500 mL
1 L
Product Type:
Amido Black Aqueous
Amido Black Methanol Base
Amido Black Methanol Rinse
Amido Black Powder
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-039-027 1 L Amido Black Aqueous
22-039-023 1 L Amido Black Methanol Base
22-039-025 1 L Amido Black Methanol Rinse
22-039-021 25 g Amido Black Powder
22-039-022 500 mL Amido Black Methanol Base
22-039-024 500 mL Amido Black Methanol Rinse
22-039-026 500 mL Amido Black Aqueous
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22-039-027 Supplier Mopec Supplier No. FS768
Add to Cart
Add to Cart
It is a forensic reagent used to enhance latent fingerprints contaminated with blood by reacting with proteins in the residue.
  • Performance
    • Enhances blood-contaminated latent prints
    • Reacts with proteins for strong, visible contrast
    • Produces blue-black staining for clear detail
    • Works on both porous and nonporous surfaces
    • Not effective on normal (non-blood) latent prints
Specifications
Product Type Amido Black Aqueous
Color Black
Quantity 1 L