Learn More
Bioworld Amido Black, 100 g
Catalog No. 50199188
Encompass_Preferred
Stain for proteins and total carbohydrates in plants. High Purity Grade EM (615nm, H2O) > 28,500
Catalog No. 50-199-188 Supplier Bioworld Supplier No. 217700052
Stain for proteins and total carbohydrates in plants. High Purity Grade EM (615nm, H2O) > 28,500