Learn More
Amido Black Staining Solution 2 x 250 mL, G-Biosciences

Description
- It is used in forensics samples due to its ability to stain blood proteins in fingerprints.
Specifications
Specifications
| Chemical Name or Material | Amido Black 10B |
| CAS | 1064-48-8 |
| Color | Deep Blue/Black |
| pH | 3.5 |
| Quantity | 250 mL |
| Molecular Weight (g/mol) | 616.49 |
| Grade | Research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.