Learn More
AdipoGen Amidoflumet
SDP
Supplier:  AdipoGen CDXA0212M025
Encompass_Preferred
Chemical. CAS 84466-05-7. Formula C9H7ClF3NO4S. MW 317.67. Synthetic. Trifluoromethanesulfonanilide pesticide. Acaricide and miticide with unknown mode of action. Compound can be used as analytical reference material.
Catalog No. 50-256-1792