Learn More
Apexbio Technology LLC Amidopyrine 58-15-1 50mg
SDP
Catalog No. 502240658
Encompass_Preferred
Amidopyrine is a small-molecule inhibitor targeting cyclooxygenase enzymes It is designed to inhibit these targets thereby reducing prostaglandin biosynthesis and regulating inflammation-related signaling pathways Amidopyrine exerts its biological activity primarily through cyclooxygenase inhibition which mediates both pain perception and febrile responses Based on these pharmacological properties amidopyrine holds research potential in the study of prostaglandin-mediated processes and in assessing candidate therapeutic agents for inflammatory and pain signaling pathways
This item is not returnable. View return policy
Catalog No. 50-224-0658 Supplier Apexbio Technology LLC Supplier No. B188350
Amidopyrine is a small-molecule inhibitor targeting cyclooxygenase enzymes It is designed to inhibit these targets thereby reducing prostaglandin biosynthesis and regulating inflammation-related signaling pathways Amidopyrine exerts its biological activity primarily through cyclooxygenase inhibition which mediates both pain perception and febrile responses Based on these pharmacological properties amidopyrine holds research potential in the study of prostaglandin-mediated processes and in assessing candidate therapeutic agents for inflammatory and pain signaling pathways