Learn More
Medchemexpress LLC Amifampridine (3,4-Diaminopyridine) | 54-96-6 | 99.99% | 109.13 | 10 G
SDP
Catalog No. 5000361948
Encompass_Preferred
Amifampridine (3,4-Diaminopyridine) is an orally active, potent, and cell-permeable voltage-gated potassium (Kv) channel blocker (PCB). It is effective in reversing BoNT/A intoxication and increases transmitter release from neuromuscular junctions (NMJs). It can be utilized for research into Lambert-Eaton myasthenic syndrome (LEMS).
  • Potent and cell-permeable voltage-gated potassium (Kv) channel blocker
  • Effective in reversing BoNT/A intoxication
  • Increases transmitter release from neuromuscular junctions
  • Suitable for Lambert-Eaton myasthenic syndrome (LEMS) research
Catalog No. 50-003-61948 Supplier Medchemexpress LLC Supplier No. HY1494610G
Add to Cart
Add to Cart
Amifampridine (3,4-Diaminopyridine) is an orally active, potent, and cell-permeable voltage-gated potassium (Kv) channel blocker (PCB). It is effective in reversing BoNT/A intoxication and increases transmitter release from neuromuscular junctions (NMJs). It can be utilized for research into Lambert-Eaton myasthenic syndrome (LEMS).
  • Potent and cell-permeable voltage-gated potassium (Kv) channel blocker
  • Effective in reversing BoNT/A intoxication
  • Increases transmitter release from neuromuscular junctions
  • Suitable for Lambert-Eaton myasthenic syndrome (LEMS) research