Learn More
Medchemexpress LLC Amifampridine (3,4-diaminopyridine) | 54-96-6 | 99.99% | 109.13 | 50 G
SDP
Catalog No. 5000360256
Encompass_Preferred
Amifampridine (3,4-diaminopyridine) is an orally active, potent, and cell permeable voltage-gated potassium (Kv) channel blocker. It demonstrates efficacy in reversing BoNT/A intoxication and enhances transmitter release from neuromuscular junctions. This product is suitable for research related to Lambert-Eaton myasthenic syndrome (LEMS).
  • Orally active, potent, and cell permeable voltage-gated potassium channel blocker.
  • Demonstrates efficacy in the reversal of BoNT/A intoxication.
  • Increases transmitter release from neuromuscular junctions.
  • Can be used for lambert-eaton myasthenic syndrome research.
Catalog No. 50-003-60256 Supplier Medchemexpress LLC Supplier No. HY1494650G
Add to Cart
Add to Cart
Amifampridine (3,4-diaminopyridine) is an orally active, potent, and cell permeable voltage-gated potassium (Kv) channel blocker. It demonstrates efficacy in reversing BoNT/A intoxication and enhances transmitter release from neuromuscular junctions. This product is suitable for research related to Lambert-Eaton myasthenic syndrome (LEMS).
  • Orally active, potent, and cell permeable voltage-gated potassium channel blocker.
  • Demonstrates efficacy in the reversal of BoNT/A intoxication.
  • Increases transmitter release from neuromuscular junctions.
  • Can be used for lambert-eaton myasthenic syndrome research.