Learn More
Cayman Chemical Amifostine thiol (hydrochloride), Purity: ≥90%, CAS: 14653-77-1, 10 mg
SDP
Catalog No. 502965071
Encompass_Preferred
A radioprotective agent and an active metabolite of amifostine; scavenges ABTS radicals in a cell-free assay; prevents γ-radiation-induced single- and double-stranded DNA breaks, as well as inhibits γ-radiation induced mutations at the HGPRT locus in V79-B310H lung fibroblast cells at 4 mM; reduces cis-DDP-induced mutations at the HGPRT locus in V79-B310H cells; increases survival, reduces the loss of bone marrow progenitor cells, and inhibits decreases in intestinal collagen thickness and crypt and Paneth cell density in mice exposed to total body γ-irradiation at 500 mg/kg prior to irradiation
Catalog No. 50-296-5071 Supplier Cayman Chemical Supplier No. 1861210 MG
A radioprotective agent and an active metabolite of amifostine; scavenges ABTS radicals in a cell-free assay; prevents γ-radiation-induced single- and double-stranded DNA breaks, as well as inhibits γ-radiation induced mutations at the HGPRT locus in V79-B310H lung fibroblast cells at 4 mM; reduces cis-DDP-induced mutations at the HGPRT locus in V79-B310H cells; increases survival, reduces the loss of bone marrow progenitor cells, and inhibits decreases in intestinal collagen thickness and crypt and Paneth cell density in mice exposed to total body γ-irradiation at 500 mg/kg prior to irradiation