Learn More
Medchemexpress LLC Amifostine | 20537-88-6 | 99.95% | 200 MG
SDP
Catalog No. 5000408495
Encompass_Preferred
Amifostine (WR2721) is a broad-spectrum cytoprotective and radioprotective agent that selectively protects normal tissues from damage. It is a potent inducer of HIF-α1 and p53, and protects cells by scavenging oxygen-derived free radicals.
  • Selectively protects normal tissues from radiation and chemotherapy damage
  • Potent hypoxia-inducible factor-α1 (HIF-α1) inducer
  • Potent p53 inducer
  • Protects cells by scavenging oxygen-derived free radicals
  • Reduces renal toxicity
  • Exhibits antiangiogenic action
Catalog No. 50-004-08495 Supplier Medchemexpress LLC Supplier No. HYB0639200MG
Amifostine (WR2721) is a broad-spectrum cytoprotective and radioprotective agent that selectively protects normal tissues from damage. It is a potent inducer of HIF-α1 and p53, and protects cells by scavenging oxygen-derived free radicals.
  • Selectively protects normal tissues from radiation and chemotherapy damage
  • Potent hypoxia-inducible factor-α1 (HIF-α1) inducer
  • Potent p53 inducer
  • Protects cells by scavenging oxygen-derived free radicals
  • Reduces renal toxicity
  • Exhibits antiangiogenic action