Learn More
Medchemexpress LLC Amifostine thiol | 31098-42-7 | 134.24 g·mol⁻1 | C5H14N2S | 5 MG
SDP
Catalog No. 5000223483
Encompass_Preferred
Amifostine thiol is the free thiol active metabolite of the cytoprotective prodrug amifostine. It functions as a cytoprotective and radioprotective agent and has been reported to activate p53 via a JNK-dependent signaling pathway. The material is supplied as a solid (molecular formula C5H14N2S, MW 134.24 g·mol⁻1) and is recommended to be stored at -20°C protected from light.
Catalog No. 50-002-23483 Supplier Medchemexpress LLC Supplier No. HY1378645MG
Add to Cart
Add to Cart
Amifostine thiol is the free thiol active metabolite of the cytoprotective prodrug amifostine. It functions as a cytoprotective and radioprotective agent and has been reported to activate p53 via a JNK-dependent signaling pathway. The material is supplied as a solid (molecular formula C5H14N2S, MW 134.24 g·mol⁻1) and is recommended to be stored at -20°C protected from light.