Learn More
Medchemexpress LLC Amifostine thiol dihydrochloride | 14653-77-1 | MFCD00460339 | ≥98.0% | 207.16 g/mol | C5H16Cl2N2S | 1 MG
SDP
Catalog No. 5000244894
Encompass_Preferred
Amifostine thiol dihydrochloride is the dihydrochloride salt of WR-1065, the active thiol metabolite of amifostine. It is supplied as a research reagent and analytical standard for studies of radioprotection, cytoprotection from chemotherapeutic agents, and p53/JNK-dependent signaling.
  • Protects normal tissues from toxic effects of certain cancer agents.
  • Activates p53 through a JNK-dependent signaling pathway.
  • Supplied as a dihydrochloride salt for improved handling and stability.
  • Provided in small research quantities suitable for analytical and biological studies.
Catalog No. 50-002-44894 Supplier Medchemexpress LLC Supplier No. HY1036401MG
Amifostine thiol dihydrochloride is the dihydrochloride salt of WR-1065, the active thiol metabolite of amifostine. It is supplied as a research reagent and analytical standard for studies of radioprotection, cytoprotection from chemotherapeutic agents, and p53/JNK-dependent signaling.
  • Protects normal tissues from toxic effects of certain cancer agents.
  • Activates p53 through a JNK-dependent signaling pathway.
  • Supplied as a dihydrochloride salt for improved handling and stability.
  • Provided in small research quantities suitable for analytical and biological studies.