Learn More
Medchemexpress LLC Amifostine thiol dihydrochloride | 14653-77-1 | MFCD00460339 | >98.0% | 207.16 g·mol⁻¹ | C5H16Cl2N2S | 1 ML
SDP
Catalog No. 5000244585
Encompass_Preferred
Amifostine thiol dihydrochloride (WR-1065 dihydrochloride) is the active thiol metabolite of amifostine used as a research reagent for radioprotection and cytoprotection. It is reported to protect normal tissues from the toxic effects of certain chemotherapeutic and radiotherapeutic agents and to activate p53 via a JNK-dependent signaling pathway, making it useful in studies of apoptosis, DNA damage response, and MDM2-p53 signaling.
  • Cytoprotective and radioprotective research reagent suitable for cellular assays.
  • Reported to activate p53 through a JNK-dependent signaling pathway.
  • Provided as a ready-to-use 10 mM solution (1 mL) for experimental convenience.
  • Also available in solid formats for formulation or analytical applications.
  • Useful for apoptosis, DNA damage response, and MDM2-p53 pathway studies.
Catalog No. 50-002-44585 Supplier Medchemexpress LLC Supplier No. HY10364010MM 1ML
Amifostine thiol dihydrochloride (WR-1065 dihydrochloride) is the active thiol metabolite of amifostine used as a research reagent for radioprotection and cytoprotection. It is reported to protect normal tissues from the toxic effects of certain chemotherapeutic and radiotherapeutic agents and to activate p53 via a JNK-dependent signaling pathway, making it useful in studies of apoptosis, DNA damage response, and MDM2-p53 signaling.
  • Cytoprotective and radioprotective research reagent suitable for cellular assays.
  • Reported to activate p53 through a JNK-dependent signaling pathway.
  • Provided as a ready-to-use 10 mM solution (1 mL) for experimental convenience.
  • Also available in solid formats for formulation or analytical applications.
  • Useful for apoptosis, DNA damage response, and MDM2-p53 pathway studies.