Learn More
Selleck Chemical LLC Amifostine trihydrate-10mg
SDP
Catalog No. 502389777
Encompass_Preferred
Amifostine trihydrate(WR2721 trihydrate) is the first approved radioprotective drug, used to decrease the risk of kidney problems caused by treatment with cisplatin.
Catalog No. 50-238-9777 Supplier Selleck Chemical LLC Supplier No. S429810MG
Amifostine trihydrate(WR2721 trihydrate) is the first approved radioprotective drug, used to decrease the risk of kidney problems caused by treatment with cisplatin.