Learn More
Cayman Chemical AmIfostInhydrate 100mg
SDP
Catalog No. 502972702
Encompass_Preferred
A prodrug that through dephosphorylation by alkaline phosphatase is converted to an active thiol, WR 1065 that has free radical scavenging, DNA protective, and hypoxia inducing activities; used to selectively reduce toxicity in normal tissues exposed to radiation or chemotherapeutic agents
Catalog No. 50-297-2702 Supplier Cayman Chemical Supplier No. 14398100 MG
A prodrug that through dephosphorylation by alkaline phosphatase is converted to an active thiol, WR 1065 that has free radical scavenging, DNA protective, and hypoxia inducing activities; used to selectively reduce toxicity in normal tissues exposed to radiation or chemotherapeutic agents