Learn More
Cayman Chemical Amifostine (hydrate), Purity: ≥95%, CAS: 112901-68-5, 50 mg
SDP
Catalog No. 502969089
Encompass_Preferred
A prodrug that through dephosphorylation by alkaline phosphatase is converted to an active thiol, WR 1065 that has free radical scavenging, DNA protective, and hypoxia inducing activities; used to selectively reduce toxicity in normal tissues exposed to radiation or chemotherapeutic agents
Catalog No. 50-296-9089 Supplier Cayman Chemical Supplier No. 1439850 MG
A prodrug that through dephosphorylation by alkaline phosphatase is converted to an active thiol, WR 1065 that has free radical scavenging, DNA protective, and hypoxia inducing activities; used to selectively reduce toxicity in normal tissues exposed to radiation or chemotherapeutic agents