Learn More
Aviva Systems Biology Corp AMIGO3 Antibody - N-terminal region (ARP55919_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55919_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AMIGO3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AMIGO3. Alternate names: MGC120552, AMIGO-3ApplicationWB. Species reactivity: Guinea Pig, Human
Catalog No. 50-014-61663