Learn More
Apexbio Technology LLC Amikacin 37517-28-5 1g
SDP
Catalog No. 5000094328
Encompass_Preferred
Amikacin (CAS 37517-28-5) is a small-molecule inhibitor targeting the bacterial 30S ribosomal subunit It is designed to interfere with protein synthesis thereby disrupting bacterial growth Amikacin exerts its biological activity primarily through specific binding to the 30S ribosomal subunit leading to inhibition of bacterial protein synthesis In in vitro studies Amikacin demonstrates antibacterial activity with reported IC50 values ranging from approximately 2 to 16 g/mL depending on bacterial species strain variations and assay conditions Based on these pharmacological properties Amikacin holds research potential in bacterial resistance studies antibiotic pharmacodynamics and antimicrobial susceptibility testing
Catalog No. 50-000-94328 Supplier Apexbio Technology LLC Supplier No. B34311000
Amikacin (CAS 37517-28-5) is a small-molecule inhibitor targeting the bacterial 30S ribosomal subunit It is designed to interfere with protein synthesis thereby disrupting bacterial growth Amikacin exerts its biological activity primarily through specific binding to the 30S ribosomal subunit leading to inhibition of bacterial protein synthesis In in vitro studies Amikacin demonstrates antibacterial activity with reported IC50 values ranging from approximately 2 to 16 g/mL depending on bacterial species strain variations and assay conditions Based on these pharmacological properties Amikacin holds research potential in bacterial resistance studies antibiotic pharmacodynamics and antimicrobial susceptibility testing