Learn More
Selleck Chemical LLC Amikacin disulfate S3065-200mg
SDP
Catalog No. 503108233
Encompass_Preferred
Amikacin sulfate (BB-K8) binds to 16S rRNA (bacterial 30S ribosome) causing misreading of mRNA and supressing proteins synthesis
Catalog No. 50-310-8233 Supplier Selleck Chemical LLC Supplier No. S3065200MG
Amikacin sulfate (BB-K8) binds to 16S rRNA (bacterial 30S ribosome) causing misreading of mRNA and supressing proteins synthesis