Learn More
Research Products International Corp Amikacin Disulfate, 25 Grams
SDP
Catalog No. 501368046
Encompass_Preferred
Amikacin disulfate is an antibiotic that belongs to the aminoglycoside class. It is commonly used in microbiology and cell culture applications. This antibiotic is a broad-spectrum aminoglycoside antibiotic with potent antibacterial activity against a wide range of Gram-negative and some Gram-positive bacteria. 
Used in microbiology experiments to inhibit the growth of bacteria in cell cultures or to select for cells containing antibiotic resistance genes. It is particularly effective against certain strains of bacteria that are resistant to other antibiotics.  
WARNING! This product can expose you to Amikacin disulfate, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-136-8046 Supplier Research Products International Corp Supplier No. A2054025.0
Amikacin disulfate is an antibiotic that belongs to the aminoglycoside class. It is commonly used in microbiology and cell culture applications. This antibiotic is a broad-spectrum aminoglycoside antibiotic with potent antibacterial activity against a wide range of Gram-negative and some Gram-positive bacteria. 
Used in microbiology experiments to inhibit the growth of bacteria in cell cultures or to select for cells containing antibiotic resistance genes. It is particularly effective against certain strains of bacteria that are resistant to other antibiotics.  
WARNING! This product can expose you to Amikacin disulfate, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.