Learn More
AdipoGen Amikacin disulfate salt
SDP
Catalog No. 502561767
Encompass_Preferred
Chemical. CAS 39831-55-5. Formula C22H43N5O13 . 2H2SO4. MW 781.76. Amikacin is a broad-spectrum aminoglycoside antibiotic and a semisynthetic analog of kanamycin. It irreversibly binds to 16S rRNA and the RNA-binding S12 protein of the 30S subunit and 50S subunit of prokaryotic ribosome and inhibits protein synthesis. It works in a concentration-dependent manner, and has better action in an alkaline environment. Amikacin disulfate is very active against most Gram-negative bacteria including gentamicin- and tobramycin-resistant strains, due to its resistance to inactivating enzymes. Amikacin disulfate also inhibits the infections caused by susceptible Nocardia and nontuberculous mycobacteria. Amikacin can be used to treat non-tubercular mycobacterial infections and tuberculosis when first-line drugs fail to control the infection.
Catalog No. 50-256-1767 Supplier AdipoGen Supplier No. CDXA0286G005
Chemical. CAS 39831-55-5. Formula C22H43N5O13 . 2H2SO4. MW 781.76. Amikacin is a broad-spectrum aminoglycoside antibiotic and a semisynthetic analog of kanamycin. It irreversibly binds to 16S rRNA and the RNA-binding S12 protein of the 30S subunit and 50S subunit of prokaryotic ribosome and inhibits protein synthesis. It works in a concentration-dependent manner, and has better action in an alkaline environment. Amikacin disulfate is very active against most Gram-negative bacteria including gentamicin- and tobramycin-resistant strains, due to its resistance to inactivating enzymes. Amikacin disulfate also inhibits the infections caused by susceptible Nocardia and nontuberculous mycobacteria. Amikacin can be used to treat non-tubercular mycobacterial infections and tuberculosis when first-line drugs fail to control the infection.