Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amikacin (From Kanamycin A, White Crystalline Powder), Fisher BioReagents

Catalog No. BP26431 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP26431 1 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP26431

missing translation for 'validMainframeMsg'

  • An antibiotic that serves as a selective agent to isolate transformed bacteria
  • Amikacin functions as an indirect inhibitor of protein synthesis in prokaryotes by binding to the 30S subunit of ribosomes
  • Useful as a selective agent to isolate bacteria that have been transformed with plasmid containing the antibiotic-resistant gene
  • Intended for research purposes only; not for manufacturing or commercial use
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37517-28-5
Molecular Formula C22H43N5O13
Molecular Weight (g/mol) 585.608
InChI Key LKCWBDHBTVXHDL-RMDFUYIESA-N
Synonym Lukadin
PubChem CID 37768
ChEBI CHEBI:2637
IUPAC Name (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide
SMILES C1C(C(C(C(C1NC(=O)C(CCN)O)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(C(C(C(O3)CN)O)O)O)N

Specifications

Type Amikacin
Physical Form Solid
pH 9.5 to 11.5
Melting Point 214°C
Packaging Amber Glass
Potency > 900μg/mg
Color White
Quantity 1 g
Merck Index 15, 400
Solubility Information completely soluble in water
Formula Weight 585.61
Chemical Name or Material Amikacin, From Kanamycin A
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
TSCA Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
Recommended Storage 0° to 5°C, desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.